(R)-(-)-2-HEPTANOL 1 * 5 g
APOSOR304043-5G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals