4-(2-BROMOETHYL)ANILINE HYDROCHLORIDE 1 * 1 g
Catalog # APOSOR902728-1G
Supplier: Apollo Scientific
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
n-Octane ≥98%
Supplier: Thermo Fisher Scientific
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Pentane for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals