1% Acid Alcohol 5 litres 1 * 1 items
Catalog # LEIC3803651E
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
Reagents for chlorine determination using Lovibond® MD photometers
Supplier: Lovibond
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
SODIUM HYDROXIDE 1 * 25 g
Supplier: Merck
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
AMS CELLBANKER 2-100ML 1 * 100 ml
Supplier: MARKET SOURCE PART PROCESS
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
Orange II ≥85 % (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Photometric test kits Robot tests
Supplier: MACHEREY-NAGEL
Reagents for Sulphide Determination using Lovibond® MD photometers
Supplier: Lovibond
IV 1000 ppm Gold for ICP in Nitric 125mL 1 * 125 mL
Supplier: MARKET SOURCE PART PROCESS
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL