2-(3,5-Dichloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA153092-250MG
Supplier: AMBEED
2-(3,5-Dichloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,98% 1 * 0,25 g
Catalog # AMBDA153092-250MG
Supplier: AMBEED
Frequently Bought Together
p-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals