1-(4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride,97% 1 * 0,25 g
Catalog # AMBDA375269-250MG
Supplier: AMBEED
1-(4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-N,N-dimethylmethanamine hydrochloride,97% 1 * 0,25 g
Catalog # AMBDA375269-250MG
Supplier: AMBEED
Frequently Bought Together
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol, EMPLURA®, Supelco®
Supplier: Merck
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
n-Hexane, anhydrous, over molecular sieves
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals