6-Chloro-1-methyl-2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,95% 1 * 0,25 g
Catalog # AMBDA497286-250MG
Supplier: AMBEED
6-Chloro-1-methyl-2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole,95% 1 * 0,25 g
Catalog # AMBDA497286-250MG
Supplier: AMBEED
Frequently Bought Together
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Methanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Chloroform stabilised for HPLC
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
5L Distilled Water 1 * 5 L
Supplier: REAGENT
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone 99.9% (by GC, corrected for water content), ECD tested halocarbons free grade for residue analysis
Supplier: Thermo Fisher Scientific