2-(3,5-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 0,1 g
Catalog # AMBDA256535-100MG
Supplier: AMBEED
2-(3,5-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,95% 1 * 0,1 g
Catalog # AMBDA256535-100MG
Supplier: AMBEED
Frequently Bought Together
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Chlorobenzene, Sigma-Aldrich®
Supplier: Merck
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.6%, Laboratory Reagent
Supplier: Honeywell Chemicals
Liquid for manometer with sloping tube
Supplier: VWR Chemicals