Methyl 2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,95+% 1 * 0,1 g
AMBDA1543055-100MG
: AMBEED
Methyl 2,4-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate,95+% 1 * 0,1 g
AMBDA1543055-100MG
: AMBEED
Frequently Bought Together
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
2,2,4-Trimethylpentane, extra dry over Molecular Sieve 99.5%, AcroSeal®
: Thermo Fisher Scientific
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
: PanReac AppliChem
Tetrachloroethylene 99%, extra pure
: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
: Thermo Fisher Scientific
n-Hexane
: Roth Carl
Cyclohexane 99.8% for HPLC
: Thermo Fisher Scientific