(2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol,98% 1 * 25 g
Catalog # AMBDA393785-25G
Supplier: AMBEED
(2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol,98% 1 * 25 g
Catalog # AMBDA393785-25G
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Pentane, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals