4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one,97% 1 * 5 g
AMBDA106243-5G
: AMBEED
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one,97% 1 * 5 g
AMBDA106243-5G
: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
MAKE-UP 1 * 750 mL
: MARKET SOURCE PART PROCESS
Water, distilled
: DISTRIBUTOR
n-Heptane 99+% for HPLC
: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
: Thermo Fisher Scientific
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
: Roth Carl
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific