(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((2,2,10,10-tetramethyl-4,8-dioxo-3,9-dioxa-5,7-diazaundecan-6-ylidene)amino)but anoic acid,97% 1 * 0,1 g
AMBDA370121-100MG
: AMBEED
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((2,2,10,10-tetramethyl-4,8-dioxo-3,9-dioxa-5,7-diazaundecan-6-ylidene)amino)but anoic acid,97% 1 * 0,1 g
AMBDA370121-100MG
: AMBEED
Frequently Bought Together
Ethylene glycol ≥99.7%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Heptane (mixture of isomers), TECHNICAL
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
n-Pentane ≥95%, TECHNICAL
: VWR Chemicals