N,N,N-Trimethyl-2,3-bis((Z)-octadec-9-en-1-yloxy)propan-1-aminium chloride,95% 1 * 0,5 g
AMBDA776776-50MG
: AMBEED
N,N,N-Trimethyl-2,3-bis((Z)-octadec-9-en-1-yloxy)propan-1-aminium chloride,95% 1 * 0,5 g
AMBDA776776-50MG
: AMBEED
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Methyl ethyl ketone ≥99%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals