3-Isopropyl-1H-pyrazole-5-carboxylic acid,97% 1 * 0,25 g
Catalog # AMBDA959017-250MG
Supplier: AMBEED
3-Isopropyl-1H-pyrazole-5-carboxylic acid,97% 1 * 0,25 g
Catalog # AMBDA959017-250MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Iodomethane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND