3-Iodo-6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole,97% 1 * 0,1 g
Catalog # AMBDA312056-100MG
Supplier: AMBEED
3-Iodo-6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole,97% 1 * 0,1 g
Catalog # AMBDA312056-100MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Supplier: Merck
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Supplier: Honeywell Chemicals
CHLOROFORM CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Water, TraceSELECT™ for trace analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific