1-(Bromomethyl)-4-(trifluoromethyl)benzene,98% 1 * 5 g
Catalog # AMBDA439908-5G
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol USP, J.T.Baker®
Supplier: Avantor
n-Hexane
Supplier: Roth Carl
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
Benzyl alcohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99%
Supplier: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
Supplier: Honeywell Chemicals