Benzyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate,98% 1 * 0,25 g
Catalog # AMBDA143850-250MG
Supplier: AMBEED
Benzyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate,98% 1 * 0,25 g
Catalog # AMBDA143850-250MG
Supplier: AMBEED
Frequently Bought Together
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
o-Xylene anhydrous, 97 % 1 * 1 L
Supplier: Merck
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
BENZYL ALCOHOL BIOTECH REAGENT 1 * 1 L
Supplier: Avantor
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals