(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol,98% 1 * 1 g
Catalog # AMBDA190057-1G
Supplier: AMBEED
(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol,98% 1 * 1 g
Catalog # AMBDA190057-1G
Supplier: AMBEED
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol 20% (w/w), Emprove® Expert for cleaning biochromatography resins, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
n-Hexane, extra dry 96+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Isopentane pure 1 * 1.000 mL
Supplier: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Super-DHB ≥99.0% matrix substance for MALDI, Supelco®
Supplier: Merck
Cyclohexane ≥99.5% (by GC), ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Dimethyl sulphoxide ≥99.9% for HPLC
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Dichloromethane
Supplier: Roth Carl
n-Hexane ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals