(4-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA390782-250MG
Supplier: AMBEED
(4-((2-Isopropyl-5-methylphenoxy)methyl)phenyl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA390782-250MG
Supplier: AMBEED
Frequently Bought Together
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Iodomethane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
1,2-Dichloroethane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND