(2S,4S)-tert-Butyl 4-amino-2-(hydroxymethyl)pyrrolidine-1-carboxylate hydrochloride,98% 1 * 0,1 g
Catalog # AMBDA728364-100MG
Supplier: AMBEED
(2S,4S)-tert-Butyl 4-amino-2-(hydroxymethyl)pyrrolidine-1-carboxylate hydrochloride,98% 1 * 0,1 g
Catalog # AMBDA728364-100MG
Supplier: AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
m-Xylene, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol Absolute 99.9% Bp Grade USP200 proof 500ml (No Auth Req) 1 * 500 mL
Supplier: DISTRIBUTOR
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Methanol bio-based ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99%
Supplier: Thermo Fisher Scientific
n-Heptane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Toluene, extra dry 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals