2,5-Dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,98% 1 * 0,25 g
Catalog # AMBDA801156-250MG
Supplier: AMBEED
2,5-Dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,98% 1 * 0,25 g
Catalog # AMBDA801156-250MG
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
n-Octane ≥98%
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Acetic acid 99-100%
Supplier: Roth Carl
Xylene (mixture of isomers) for histology
Supplier: Honeywell Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific