(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid,97% 1 * 0,25 g
Catalog # AMBDA509784-250MG
Supplier: AMBEED
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanecarboxylic acid,97% 1 * 0,25 g
Catalog # AMBDA509784-250MG
Supplier: AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
9-Fluorenylmethyl chloroformate (Fmoc chloride), Supelco®
Supplier: Merck
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether 99%, pure
Supplier: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals