3-(Trifluoromethyl)aniline hydrochloride,98% 1 * 100 g
Catalog # AMBDA750507-100G
Supplier: AMBEED
Frequently Bought Together
Chloroform, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Diethylene glycol for synthesis
Supplier: Merck
2-Butoxyethanol 99%, extra pure
Supplier: Thermo Fisher Scientific
Chloroform, extra dry 99.9% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
GLYCEROL PURISS., MEETS ANALYTICAL SPECI 1 * 1 L
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute for histology, J.T.Baker®
Supplier: Avantor
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Chlorobenzene, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone 99.6% ACS
Supplier: Thermo Fisher Scientific
CYCLOPENTANE REAGENT GRADE 98% 1 * 500 mL
Supplier: Merck
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific