1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione,98% 1 * 25 g
Catalog # AMBDA133207-25G
Supplier: AMBEED
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione,98% 1 * 25 g
Catalog # AMBDA133207-25G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific