4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol),97% 1 * 25 g
Catalog # AMBDA361290-25G
Supplier: AMBEED
4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol),97% 1 * 25 g
Catalog # AMBDA361290-25G
Supplier: AMBEED
Frequently Bought Together
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
4-Methyl-2-pentanone ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
ETHANOL ABSOLUTE BAKER ANALYZED 1 * 25 L
Supplier: Avantor
tert-Butanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethylene glycol monobutyl ether ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), extra dry 98+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for HPLC
Supplier: VWR Chemicals