2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-8-(tert-butoxycarbonyl)-2,8-diazaspiro[4.5]decane-3-carboxylic acid,95% 1 * 0,1 g
Catalog # AMBDA213727-100MG
Supplier: AMBEED
2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-8-(tert-butoxycarbonyl)-2,8-diazaspiro[4.5]decane-3-carboxylic acid,95% 1 * 0,1 g
Catalog # AMBDA213727-100MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
n-Pentane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Nonane 99%, Reagent Grade
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetonitrile 99.9% ACS, gradient grade, meets analytical specification of Ph.Eur. for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,4-Dioxane ≥99% stabilised
Supplier: Thermo Fisher Scientific
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals
Dichloromethane ≥99% (by GC) stabilised, Puriss., meets analytical specification of Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals