3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate,99+% 1 * 0,25 g
Catalog # AMBDA782133-250MG
Supplier: AMBEED
3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate,99+% 1 * 0,25 g
Catalog # AMBDA782133-250MG
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Triethylene glycol dimethyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
5L Distilled Water 1 * 5 L
Supplier: REAGENT
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific