(S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexano ic acid,97% 1 * 5 g
Catalog # AMBDA178274-5G
Supplier: AMBEED
(S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-6-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexano ic acid,97% 1 * 5 g
Catalog # AMBDA178274-5G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Benzoic acid methyl ester
Supplier: Merck
Diethyl ether BHT 1ppm inhibitor, anhydrous, 99.7 % 1 * 1 L
Supplier: Merck
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
N-Ethyldiisopropylamine 99.5+%
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Diethylene glycol monobutyl ether ≥99%
Supplier: Thermo Fisher Scientific
n-Heptane ∼ 99% (n-heptane) for HPLC
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
Supplier: Thermo Fisher Scientific
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals