(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid,95% 1 * 5 g
Catalog # AMBDA121129-5G
Supplier: AMBEED
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid,95% 1 * 5 g
Catalog # AMBDA121129-5G
Supplier: AMBEED
Frequently Bought Together
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol for HPLC
Supplier: Thermo Fisher Scientific
Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 1 L
Supplier: PanReac AppliChem
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Cesium fluoride 99.9%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem