1-Cyclopropyl-6-fluoro-7-((1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid methanes ulfonic acid salt,98% 1 * 5 g
Catalog # AMBDA302286-5G
Supplier: AMBEED
1-Cyclopropyl-6-fluoro-7-((1S,4S)-5-methyl-2,5-diazabicyclo[2.2.1]heptan-2-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid methanes ulfonic acid salt,98% 1 * 5 g
Catalog # AMBDA302286-5G
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethyl acetate, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific