N2-(tert-Butyl)-N4-cyclopropyl-6-(methylthio)-1,3,5-triazine-2,4-diamine,95% 1 * 25 g
Catalog # AMBDA402724-25G
Supplier: AMBEED
N2-(tert-Butyl)-N4-cyclopropyl-6-(methylthio)-1,3,5-triazine-2,4-diamine,95% 1 * 25 g
Catalog # AMBDA402724-25G
Supplier: AMBEED
Frequently Bought Together
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetramethylammonium hydrogen sulphate, Supelco®
Supplier: Merck
1,4-Dioxane ≥99.5% (by GC) stabilised, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diisopropyl ether, Sigma-Aldrich®
Supplier: Merck
Petroleum spirit, 50…70 °C ≥90%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Ethanol (untaxed) ≥99.8%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals