Ethyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-5-carboxylate,95% 1 * 0,1 g
AMBDA1669302-100MG
: AMBEED
Ethyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-5-carboxylate,95% 1 * 0,1 g
AMBDA1669302-100MG
: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Ethyl acetate ≥99%, TECHNICAL
: VWR Chemicals
Toluene ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
: MERCK PRODUCTION CHEMICALS
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Glycerine, Sigma-Aldrich®
: Merck
Benzyl alcohol ≥99%
: Thermo Fisher Scientific
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals