4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol hydrobromide,97% 1 * 0,25 g
Catalog # AMBDA1372882-250MG
Supplier: AMBEED
4-((2,4-Diaminopyrimidin-5-yl)methyl)-2,6-dimethoxyphenol hydrobromide,97% 1 * 0,25 g
Catalog # AMBDA1372882-250MG
Supplier: AMBEED
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Dibutyl ether anhydrous, 99.3 % 1 * 2 L
Supplier: Merck
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Naphthol for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonedimethyl acetal for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, extra dry over Molecular Sieve 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals