3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium chloride,98% 1 * 5 g
Catalog # AMBDA990719-5G
Supplier: AMBEED
3-Carbamoyl-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyridin-1-ium chloride,98% 1 * 5 g
Catalog # AMBDA990719-5G
Supplier: AMBEED
Frequently Bought Together
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Trifluoroacetic acid, Sigma-Aldrich®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals