7-Bromo-5-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride,95% 1 * 0,1 g
Catalog # AMBDA890411-100MG
Supplier: AMBEED
7-Bromo-5-fluoro-1,2,3,4-tetrahydro-isoquinoline hydrochloride,95% 1 * 0,1 g
Catalog # AMBDA890411-100MG
Supplier: AMBEED
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
N,N-Dimethylacetamide 99.5+% for analysis
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Xylene (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
2-Methylbutane for HPLC
Supplier: Thermo Fisher Scientific
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Diphosphorus pentoxide, Supelco®
Supplier: Merck
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals