Cabinet for 24 FINA/MEGA trays - stackable 1 * 1 items
Catalog # CLLPWDP-CA-2084_1
Supplier: CELLPATH
Cabinet for 24 FINA/MEGA trays - stackable 1 * 1 items
Catalog # CLLPWDP-CA-2084_1
Supplier: CELLPATH
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Assistent Brushes for Bottles
Supplier: GLASWARENFABRIK KARL HECHT
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Blue Nitrile™
Supplier: KIMBERLY CLARK