Order Entry
United Kingdom
ContactUsLinkComponent
Multitron Standard 2 x 25 mm throw on 13cm base Temperature Range ambient +6°C to 65°C Speed range 20 – 400 rpm Power supply 230v 50Hz Pre-drilled Tray for use with clamps (Clamps not included) Double Stack dimensions: 1070w x 870d x 1195h 1 * 1 items
Catalog # INFOAN128966
Supplier:  INFORS
Multitron Standard 2 x 25 mm throw on 13cm base Temperature Range ambient +6°C to 65°C Speed range 20 – 400 rpm Power supply 230v 50Hz Pre-drilled Tray for use with clamps (Clamps not included) Double Stack dimensions: 1070w x 870d x 1195h 1 * 1 items
Catalog # INFOAN128966
Supplier:  INFORS
Frequently Bought Together