Order Entry
United Kingdom
ContactUsLinkComponent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Fraction Collector. LC System Compatibilit y: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. Technique: LC & LC/MS LC. 1 * 1 items
Catalog # HEWLG7158-68100
Supplier:  Agilent
Specifications:- LC Application Compatibility: Preparative. LC Module Type Compatibility: Fraction Collector. LC System Compatibilit y: 1290 Infinity II Purification system. LC Use With Instrument: G7158B G7159B. Technique: LC & LC/MS LC. 1 * 1 items
Catalog # HEWLG7158-68100
Supplier:  Agilent
Frequently Bought Together