Order Entry
United Kingdom
ContactUsLinkComponent
UV instrument USP/EP qualification set, for wavelength and photometric accuracy, stray light and resolution tests. Includes 5 and 10 mm pathlength cuvettes to allow stray light testing using the filter ratio method outlined in USP Chapter 857. Suppl 1 * 1 items
Catalog # HEWLG9864-80009
Supplier:  Agilent
UV instrument USP/EP qualification set, for wavelength and photometric accuracy, stray light and resolution tests. Includes 5 and 10 mm pathlength cuvettes to allow stray light testing using the filter ratio method outlined in USP Chapter 857. Suppl 1 * 1 items
Catalog # HEWLG9864-80009
Supplier:  Agilent
Frequently Bought Together