Order Entry
United Kingdom
ContactUsLinkComponent
Specifications:- Cone Care Package: Yes. Interface Cone Base Material: Nickel-plated Copper. Interface Cone Tip Material: Platinum. Spectroscopy Instrument Compatibility: 7700 7800 7900 8800 8900 7850. 1 * 1 items
Catalog # HEWL5067-0297
Supplier:  Agilent
undefined
Specifications:- Cone Care Package: Yes. Interface Cone Base Material: Nickel-plated Copper. Interface Cone Tip Material: Platinum. Spectroscopy Instrument Compatibility: 7700 7800 7900 8800 8900 7850. 1 * 1 items
Catalog # HEWL5067-0297
Supplier:  Agilent
Frequently Bought Together