Order Entry
United Kingdom
ContactUsLinkComponent
Specifications:- Cone Care Package: Yes. Interface Cone Base Material: Copper. Interface Cone Tip Material: Nickel. Spectroscopy Ins trument Compatibility: 7700 7800 7900 8800 8900 7850. 1 * 1 items
Catalog # HEWL5067-0294
Supplier:  Agilent
undefined
Specifications:- Cone Care Package: Yes. Interface Cone Base Material: Copper. Interface Cone Tip Material: Nickel. Spectroscopy Ins trument Compatibility: 7700 7800 7900 8800 8900 7850. 1 * 1 items
Catalog # HEWL5067-0294
Supplier:  Agilent
Frequently Bought Together