ASSAYCOMPLETE CELL PLATING 38 REAGENT 1 * 500 mL
DISX93-0563R38B
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, <i>Taq</i> DNA polymerase
: VWR Chemicals
E.Z.N.A.® Blood DNA Kit
: OMEGA BIO-TEK
VWR®, dNTPs
: VWR Chemicals
Decontamination solution, RNase AWAY®
: Thermo Fisher Scientific
Serological pipettes, Stripette™
: Corning
qScript™ XLT One-Step RT-qPCR ToughMix®
: Quantabio
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
: VWR Collection
VWR® Grinding Balls for Bead Beater
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Kits for Precellys®
: Bertin Instruments
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
E.Z.N.A.® Plasmid Mini kit I
: OMEGA BIO-TEK