ASSAYCOMPLETE CELL PLATING 20 REAGENT 1 * 100 mL
Catalog # DISX93-0563R20A
Supplier: DISCOVERX
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
repliQa HiFi ToughMix
Supplier: Quantabio
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%
Supplier: VWR Chemicals
VWR®, Petri Dish, PS
Supplier: VWR Collection
E.Z.N.A.® Mollusc & Insect DNA Kit
Supplier: OMEGA BIO-TEK
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
Bioanalyzer RNA Analysis
Supplier: AGILENT
Coomassie Protein Stain InstantBlue 1 * 1.000 mL
Supplier: Abcam
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
VWR®, dNTPs
Supplier: VWR Chemicals
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
H-F ANTIDOTE GEL FIRST RESPONSE KIT 1 * 1 KIT
Supplier: INDUSTRIAL PHARMACEUTICAL SERV
CATHEPSIN B ASSAY KIT MAGIC RED 1 * 100 Tests
Supplier: Abcam
Ready DNA Ultra 2.0 prefilled plates, for KingFisher Flex instruments, KingFisher™
Supplier: Thermo Fisher Scientific
DNA ladders, GeneRuler™ and O'GeneRuler™
Supplier: Thermo Fisher Scientific