Potassium fluoride 40% (w/v) in aqueous solution, RC610016, R6100000-500A 1 * 500 mL
Catalog # RICCR6100000-500A
Supplier: RICCA CHEMICAL
Potassium fluoride 40% (w/v) in aqueous solution, RC610016, R6100000-500A 1 * 500 mL
Catalog # RICCR6100000-500A
Supplier: RICCA CHEMICAL
Frequently Bought Together
Test kits, iron, Spectroquant®, Supelco®
Supplier: Merck
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Test paper, qualitative, Watesmo
Supplier: MACHEREY-NAGEL
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Reagents for determination of silica
Supplier: Hach