Zero Waste Box™ Recycling Solution for Plastic Packaging
Product Details & Documents
A Zero WasteBox™ is a complete and convenient solution which includes the storage, shipping and recycling of a particular type of waste that isn’t currently recycled through local councils or traditional recycling facilities.
- Reduce the amount of waste that you send to landfill or incinerate
- Recycle the non-recyclable and reduce your carbon footprint
- Improving your organisation's sustainability can boost your corporate image and improve employee engagement and satifaction
Use this Zero WasteBox™ to recycle any flexible plastic packaging, carrier bags and shipping material including bubble wrap, polystyrene, airbags, shrink wrap, cushioning pallet liners and covers. Any hard/rigid plastic packaging and plastic containers. Use this box to recycle VWR® PureStep EnviroTack™ mat layers, including top and bottom release liners.
How it works:
Order - Select a Zero WasteBox™ based on what and how much you would like to recycle
Collect - Fill your box with the accepted waste and seal the box with adhesive tape
Ship - Using the pre-paid UPS shipping label affixed to the back of your box, send your filled box to TerraCycle® for recycling. UPS requires a minimum of 24 hours’ notice and does pick ups Monday to Friday between 9am and 5pm. Pick-ups can be arranged online at ups.com/gb or by telephone on 03457 877 877. All shipments are carbon-neutral, meaning the carbon emissions released during transport will be offset
Reorder - Continue to recycle and reorder a new Zero WasteBox™. Your collected waste will find a second life as recycled materials, reducing the need to extract virgin materials and depleting resources
Cardboard boxes are made from 84% recycled material. Box container and liner are 100% recyclable.
Certifications: TerraCycle® will not landfill or incinerate any of the waste we receive. An annual waste transfer note is required for waste originating from a commercial activity in the UK. Check our guide in documentation on how to fill in your TerraCycle® waste transfer note.
Caution: Do not send in food content and liquid waste. Plant-based or biodegradable plastics (e.g. PLA). Any non-plastic waste: paper, metals, cardboard, glass. Plastic cards. Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people. Batteries, pressurised canisters, broken glass and medical waste.
Documents
Specifications
- Capacity:144 L
- Description:Disposal box for plastic packaging, large
- Seq:03
- W×D×H:380×380×970 mm
- Load capacity:≤30 kg
- Environmental certification:FSC (Licence code FSC-C084464), Bureau Veritas, Third party verified LCA
- Environmentally Preferable:Product Transparency and Disclosure|Waste Reducing
- Cat. No.:129-2480
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



