Order Entry
United Kingdom
ContactUsLinkComponent
Gram BioCompact II RR410L Fridge Capacity 312L Temperature range: +2C/+20C Ambient temperature range: 10C/35C Acoustic and visual, High/Low temperature and door alarms 1 access port: 24.5 mm ABS interior lining Refrigerant R600a Supplied with 5 1 * 1 items
Catalog # GRAMRR410L
Supplier:  GRAM
Gram BioCompact II RR410L Fridge Capacity 312L Temperature range: +2C/+20C Ambient temperature range: 10C/35C Acoustic and visual, High/Low temperature and door alarms 1 access port: 24.5 mm ABS interior lining Refrigerant R600a Supplied with 5 1 * 1 items
Catalog # GRAMRR410L
Supplier:  GRAM
Frequently Bought Together