Order Entry
United Kingdom
ContactUsLinkComponent
Multitron Incubator Shaker 3 x 25mm Ø throw on low 13 base Temperature Range ambient +6C to 65C Speed range 20 – 400rpm Power supply 230V 50Hz CO2 Control 0 – 20% Hygienic Humidity Control Autoclavable reservoir for humidification Universal Tray with 1 * 1 items
Catalog # INFOAN125823
Supplier:  INFORS
Multitron Incubator Shaker 3 x 25mm Ø throw on low 13 base Temperature Range ambient +6C to 65C Speed range 20 – 400rpm Power supply 230V 50Hz CO2 Control 0 – 20% Hygienic Humidity Control Autoclavable reservoir for humidification Universal Tray with 1 * 1 items
Catalog # INFOAN125823
Supplier:  INFORS
Frequently Bought Together