Corning® Lambda™ Plus Pipettors
Product Details & Documents
Measure and transfer liquids precisely and safely using the Corning® Lambda™ Plus pipettor.
- Single-channel, variable volume, volumetric pipettors
- Universal fit for all common pipette tip brands
- Low pipetting force
- Adjustable tip ejector
- Fully autoclavable and UV-resistant
These single-channel, volumetric instruments are designed for a universal fit with all common brands of pipette tips, including Corning® Filtered pipette tips. Each pipettor features a smooth plunger movement, low pipetting force, and an adjustable tip ejector. Volume can be adjusted using the thumb wheel or plunger button, and a locking mechanism can be used to prevent accidental volume changes. This collection of pipettors can accommodate volumes from 0,2 to 1000 µl depending on the model.
Pipettors are available individually in 2, 10, 20, 200 and 1000 µl sizes or can be purchased as a kit.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Volume:0,2 - 2 µl
- Description:Lambda™ Plus pipettor
- Seq:01