Dichloro{1,2-Bis[(2R,5R)-2,5 Dimethylpho 1 * 500 mg
Catalog # STRE44-6015-500MG
Supplier: Strem Chemicals, Inc.
Dichloro{1,2-Bis[(2R,5R)-2,5 Dimethylpho 1 * 500 mg
Catalog # STRE44-6015-500MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Pentane reagent grade, 98% 1 * 1 L
Supplier: Merck
1,2-Dichloroethane, Sigma-Aldrich®
Supplier: Merck
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
N-Bromosuccinimide 99%
Supplier: Thermo Fisher Scientific
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals