Dichloro{(S)-(-)-2,2'-Bis[Di(3,5-Xylyl)P 1 * 250 mg
Catalog # STRE44-0213-250MG
Supplier: Strem Chemicals, Inc.
Dichloro{(S)-(-)-2,2'-Bis[Di(3,5-Xylyl)P 1 * 250 mg
Catalog # STRE44-0213-250MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Pentane ≥99.5% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Butyl acetate ≥96%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific