2-[(4R)-4,5-Dihydro-4-Isopropyl-2-Oxazol 1 * 100 mg
Catalog # STRE07-1049-100MG
Supplier: Strem Chemicals, Inc.
2-[(4R)-4,5-Dihydro-4-Isopropyl-2-Oxazol 1 * 100 mg
Catalog # STRE07-1049-100MG
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
TERT. BUTANOL FOR HPLC 99,5% 1 * 1 L
Supplier: Merck
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific